Learn More
Spherotech ACCUCOUNT ULTRARAINBOW CALIBRA
Catalog No. NC2023795
Encompass_Preferred
AccuCount UltraRainbow Calibration Particles, 5x106/mL, 20.0 µm (+/-2 µm), 1ml
Catalog No. NC2023795 Supplier Spherotech Supplier No. ACURFP52001
AccuCount UltraRainbow Calibration Particles, 5x106/mL, 20.0 µm (+/-2 µm), 1ml