Learn More
Rarecyte RareCyte AccuCyte Blood Collection Tube, Sterile, 10-Pack
Catalog No. NC1780451
Encompass
A pack of ten sterile AccuCyte Blood Collection Tubes (BCTs). Contents in the tubes stabilize and preserve nucleated cells in whole blood.
Catalog No. NC1780451 Supplier Rarecyte Supplier No. 241208000
A pack of ten sterile AccuCyte Blood Collection Tubes (BCTs). Contents in the tubes stabilize and preserve nucleated cells in whole blood.