Learn More
Andwin Scientific ACCUEDGE RPLCMT BLADE SCISSORS
SDP
Catalog No. 50727507
Encompass_Preferred
The Accu-Edge replacement blade scissors by Sakura Finetek feature sharp blades that also eliminate the chance of cross-contamination every time it is used for tissue processing. The blades are autoclavable and are free of corrosion. Made from stainless steel, the blades can be inserted and removed with the use of an enclosed tool with no difficulty.
Catalog No. 50-727-507 Supplier Andwin Scientific Supplier No. 6257001
The Accu-Edge replacement blade scissors by Sakura Finetek feature sharp blades that also eliminate the chance of cross-contamination every time it is used for tissue processing. The blades are autoclavable and are free of corrosion. Made from stainless steel, the blades can be inserted and removed with the use of an enclosed tool with no difficulty.