Learn More
Andwin Scientific ACCUEDGE SCISSORS
SDP
Catalog No. NC9388746
Encompass
The Tissue-Tek Accu-Edge replaceable blade scissors are ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold individually.
Catalog No. NC9388746 Supplier Andwin Scientific Supplier No. 25609012
The Tissue-Tek Accu-Edge replaceable blade scissors are ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold individually.