Learn More
ACCUTEC BLADES INC (3P) AccuForge Double Edge Blade with Microcoat 004" Stainless Steel Blades, Wrapped, 250 Blades per Carton, 20 Cartons per Case
Catalog No. NC3721102
Encompass_Preferred
AccuForge Double Edge blades feature a 3-Facet edge for exacting, clean cuts. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Blades are individually wrapped for safety. Made in the USA
Catalog No. NC3721102 Supplier ACCUTEC BLADES INC (3P) Supplier No. AGBL60000000
AccuForge Double Edge blades feature a 3-Facet edge for exacting, clean cuts. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Blades are individually wrapped for safety. Made in the USA