Learn More
ACCUTEC BLADES INC (3P) AccuForge GEM STAR Scientific Single Edge Blade Dispenser SKU# AGBL-7008-0000 .009" Carbon Steel 3-Facet Blades, Aluminum Back, 10 Blades per Dispenser, 12 Dispensers per Carton, 12 Cartons per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70080000
Encompass_Preferred
AccuForge GEM STAR Scientific single edge blades feature .009 carbon steel and 3-facet edges for sharpness. Each dispenser holds 10 blades and includes used blade storage. Units per Case: 144
Catalog No. NC3721115