Learn More
ACCUTEC BLADES INC (3P) AccuForge GEM3 Scientific Single Edge Blade Cartridge SKU# AGBL-7011-0000 .009" Carbon Steel 3-Facet Blades, Steel Back, 100 Blades per Cartridge, 10 Cartridges per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70110000
Encompass_Preferred
AccuForge Gem Scientific Single Edge blades feature .009 high carbon steel for durability, and 3-facet edges for sharpness. Each cartridge contains 100 blades. Fits Gem Runner™ Dispenser AGTL-7000-0000. Units per Case: 10
Catalog No. NC3721124