Learn More
ACCUTEC BLADES INC (3P) AccuForge Single Edge Long Blade SKU# AGBL-7002-0000 .009" Stainless Steel 3-Facet Unbacked Blades, 500 Blades per Case
Catalog No. NC3721112
Encompass_Preferred
AccuForge Single Edge Long blades feature a 2.25" long length for extra cutting width and 3-facet edges for increased sharpness. Blades are unbacked and uncoated. Bulk pack of 500 blades.
Catalog No. NC3721112 Supplier ACCUTEC BLADES INC (3P) Supplier No. AGBL70020000
AccuForge Single Edge Long blades feature a 2.25" long length for extra cutting width and 3-facet edges for increased sharpness. Blades are unbacked and uncoated. Bulk pack of 500 blades.