Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Inventory and Floor Marking Tape

Catalog No. 19805174 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-174 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-174 Supplier Accuform Signs Supplier No. PTM636YL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solid color marking tape increases pipe visibility and helps to identify hazardous areas.

  • 5-mil tape with self-adhesive, no peel-off liner
  • Well-suited for a multitude of uses
  • Adhere to floors, walls, piping, posts, beams, handrails, stairways, machinery and equipment
  • Waterproof and wear-resistant, with minor resistance to acids, oils, grease, caustics, alkalis and most solvents
  • 3 in. x 180 ft.
  • Service temperature range: -20° to 140°F (-29° to 60°C)

Specifications

Color Yellow
Dimensions (L x W) 180 ft. x 3 in.
Material Vinyl
Thickness 6 mil
Temperature Range (Metric) −28.9°C to 82.2°C
Width (English) 3 in.
Width (Metric) 7.6 cm
For Use With (Application) Identify hazardous areas, Mark walkways or Use in 5S/Lean initiatives
Length (English) 180 ft.
Length (Metric) 54.86 m
Type Solid-colored Tape
Temperature Range (English) −20°F to 140°F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.