Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Barricade Tapes

Catalog No. 19804923 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black, Red
Black, Yellow
Red, Black
Displayed Text:
Danger Asbestos Hazard
Danger Keep Out
Police Line Do Not Cross
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Displayed Text
19-804-923 Black, Red Danger Asbestos Hazard
19-804-925 Black, Yellow Police Line Do Not Cross
19-804-926 Red, Black Danger Keep Out
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-804-923 Supplier Accuform Signs Supplier No. MPT05
Add to Cart
Add to Cart

High-visibility barricade tapes are a quick, temporary and effective way to surround or section-off hazard areas, danger zones and construction sites

  • Size: 7.6cm x 304.8m (3 in. x 1000 ft.)
  • Material: Polyethylene plastic
  • Non-adhesive
  • Self-wound on a roll
  • Unspool, cut easily, and can be fastened with any method
  • Clear, bold warning message that is continuously repeated the length of the roll
  • Material is economical and disposable, yet can be reused

Specifications

Color Black, Red
Width (Metric) 7.62 cm
Length (Metric) 304.8 m
Thickness 3 mil
Width (English) 3 in.
Length (English) 1000 ft.
Adhesive Type Non-adhesive
Certifications/Compliance ANSI Z535.5-3.2
For Use With (Application) Crowd control
Material HDPE
Type Barricade Tape
Dimensions (L x W) 1000 ft. x 3 in. (304.8 m x 7.62 cm)
Displayed Text Danger Asbestos Hazard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.