Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Calibration Labels

Catalog No. 19809812 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-809-812 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-809-812 Supplier Accuform Signs Supplier No. LPC422
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Calibration labels can be used to inform and protect workers and employees.

Specifications

Certifications/Compliance OSHA 29 CFR 1910.145, ANSI Z535
Material Vinyl
Quantity 25/Pk.
Adhesive Type Acrylic
Mounting Type Surface Mount
For Use With (Application) To identify the status of a given item or product generally with the date and signature of person declaring the status
Thickness 4 mil
Width (English) 1.5 in.
Length (English) 0.62 in.
Length (Metric) 1.57 cm
Product Type Calibration Label
Color White, Green
Life Span 2 to 4 Years
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.