Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Caution Do Not Enter Barricade Tape

Catalog No. 19821810 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black, Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-821-810 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-821-810 Supplier Accuform Signs Supplier No. MPT31
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-visibility barricade tapes are a quick, temporary, effective way to section-off hazard areas, danger zones and construction sites

  • Label: CAUTION. DO NOT ENTER
  • Size: 7.6cm x 304.8m (3 in. x 1000 ft.)
  • Material: Polyethylene plastic
  • Non-adhesive
  • Self-wound on a roll
  • Unspool, cut easily and can be fastened with any method
  • Clear, bold warning messge that is continuously repeated the length of the roll
  • Color: Yellow, Black

Specifications

Color Black, Yellow
Width (Metric) 7.6 cm
Width (English) 3 in.
Length (English) 1000 ft.
For Use With (Application) Crowd control
Length (Metric) 304.8 m
Thickness 3 mil
Displayed Text Caution Do Not Enter
Adhesive Type Non-adhesive
Material HDPE
Type Barricade Tape
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.