Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Chain Links

Catalog No. 19805122 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-122 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-122 Supplier Accuform Signs Supplier No. PRC202YL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All-plastic chain will not rust or corrode

Polyethylene plastic chain links measure 2 x 1 x 1/4 in.. Used with stanchion posts for crowd control and to guide pedestrian traffic.

Order Info

Available in white, yellow, black, or red. Sold by the foot.

Specifications

Color Yellow
For Use With (Application) Used with stanchion posts for crowd control and to guide pedestrian traffic
Material Polyethylene
Type Strong Chain Link
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.