Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Chemicals and Haz Mat Signs: Asbestos

Catalog No. 19804660 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
7 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-804-660 7 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-660 Supplier Accuform Signs Supplier No. MCAW101VS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inform all readers of a cancer and lung disease hazard brought about by asbestos.

  • Legend: DANGER ASBESTOS CANCER AND LUNG DISEASE HAZARD AUTHORIZED PERSONNEL ONLY
  • Size: 25.4 × 35.6cm (10 × 14 in.)
  • Material: Adhesive vinyl
  • Color: White, Black, Red
  • Language: English

This sign is being phased out by June 1, 2016 and replaced with update verbiage.

Specifications

Material Vinyl
Quantity 1 Ea.
Height (English) 10 in.
Height (Metric) 25.4 cm
Max. Temperature (English) 176°F
Min. Temperature (English) -20°F
Width (English) 14 in.
Width (Metric) 35.6 cm
Temperature Range (Metric) -29°C to 80°C
Temperature Range (English) -20°F to 176°F
Dimensions (L x W) 7 x 10 in.
Displayed Text DANGER Asbestos Cancer And Lung Disease Hazard Authorized Personnel Only
Product Type Safety Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.