Learn More
Accuform Signs Chilled Water Supply Pipe Markers
Catalog No. 19804875 Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Chilled Water Supply
Catalog No. Display Text
19-804-875 Chilled Water Supply
1 options
Catalog No. 19-804-875 Supplier Accuform Signs Supplier No. RPK203SSD
Add to Cart
Add to Cart
Identify contents and mark the directional flow for pipes used for chilled water supply.
Specifications
Description Pipe Marker
Adhesive Type Self-adhesive
Material Vinyl, Plastic
Display Text Chilled Water Supply
For Use With (Application) Clearly identifying the pipe contents and flow direction, make maintaining facility easier and increase employee safety by clearly marking pipes
Life Span 1 to 3 Years
Quantity 1 Ea.
Temperature Range (English) -90 to 168°F
Temperature Range (Metric) -67.77 to 75.55°C
Thickness (English) 4 mil
Diameter (English) 2.5 to 6 in.
Diameter (Metric) 6.35 to 15.24 cm