Learn More
Description
- Legend: DANGER. CONFINED SPACE. DO NOT ENTER
- Available in a variety of sizes and materials
- Color: White, Black, Red
- Language: English
Specifications
Specifications
| Certifications/Compliance | 29 CFR 191.146 |
| Material | Vinyl |
| Quantity | 1 Ea. |
| For Use With (Application) | Intended to indicate potential, type and degree of the hazard that may lead to accidental injury or property damage |
| Height (English) | 10 in. |
| Height (Metric) | 25.4 cm |
| Max. Temperature (English) | 146°F |
| Min. Temperature (English) | -20°F |
| Thickness | 4 mil |
| Width (English) | 14 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.