Learn More
Accuform Signs Edge Sealer
Catalog No. 19804654 Shop All AccuformNMC Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-804-654 1 Ea.
1 options
Catalog No. 19-804-654 Supplier Accuform Signs Supplier No. MB144
Add to Cart
Add to Cart
Add extra adhesion power quickly with this edge sealer that fastens things together.
Specifications
Product Type Edge Sealer
Format Viscous Liquid
For Use With (Application) Fastens signs and tapes to floors, walls, drums or containers, prevents edges from lifting and inhibits chemical intrusion
Adhesive Type Self-adhesive
Quantity 1 Ea.
Volume (English) 5 oz.
Volume (Metric) 141.74 g