Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Electronic Moving Signs with LED

Catalog No. 19804843 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
49 x 4.25 x 6.25 in. (LxDxH)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-804-843 49 x 4.25 x 6.25 in. (LxDxH)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-843 Supplier Accuform Signs Supplier No. MME246
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Get maximum visual impact from personnel with attention-grabbing messages.

Specifications

Includes Hardware
Quantity 1 Ea.
For Use With (Application) An electronic moving message display attracts attention to greetings and safety alerts
Height (English) 6.25 in.
Width (English) 49 in.
Dimensions (L x W) 49 x 4.25 x 6.25 in. (LxDxH)
Displayed Text Electronic Moving Message Display
Product Type Electronic Message Board
Color Multi-colored
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.