Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Hazrds Signs: Danger: Flammable

Catalog No. 17998062 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Vinyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
17-998-062 Vinyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-998-062 Supplier Accuform Signs Supplier No. MCHL231VS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

OSHA-compliant sign warns of potential flammable materials in the work environment.

Specifications

Material Vinyl
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Indoor and outdoor use
Height (English) 10 in.
Max. Temperature (English) 146°F
Min. Temperature (English) -20°F
Thickness 4 mil
Width (English) 14 in.
Temperature Range (Metric) -29°C to 63°C
Temperature Range (English) -20°F to 146°F
Displayed Text DANGER Flammable
Product Type Hazard Sign
Color Red on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.