Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Marking Paints

Catalog No. 19804454 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Fluorescent Orange
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-804-454 Yellow
19-821-767 Fluorescent Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-804-454 Supplier Accuform Signs Supplier No. FMP229

Mark sites and locations as well as stripe underground lines, aisles, pathways, parking lines and traffic lines

  • Solvent based
  • Can be applied to virtually any indoor and outdoor surface (concrete, asphalt, gravel, soil and grass)
  • 17 oz. cans are designed for upside-down use with non-clogging spray tips
  • Contains fast-drying, long-lasting, high-solids formula paint that penetrates and adheres quickly to most surfaces
  • Lead-free
  • Available in a variety of colors

Not available for sale in Canada

Order Info

Due to DOT regulations this item must ship via ground service only. It is not eligible to ship overnight, 2-day or 3-day.

Compliance

VOC compliant

Specifications

Quantity 1 Ea.
Capacity (English) 17 oz.
Product Type Paint
For Use With (Application) Solvent based Utility, Mark Sites and Stripe Underground Lines, Aisles, Pathways, Parking, Traffic Lines
Certifications/Compliance VOC
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.