Learn More
Description
- Solvent based
- Can be applied to virtually any indoor and outdoor surface (concrete, asphalt, gravel, soil and grass)
- 17 oz. cans are designed for upside-down use with non-clogging spray tips
- Contains fast-drying, long-lasting, high-solids formula paint that penetrates and adheres quickly to most surfaces
- Lead-free
- Available in a variety of colors
Not available for sale in Canada
Order Info
Due to DOT regulations this item must ship via ground service only. It is not eligible to ship overnight, 2-day or 3-day.
Compliance
VOC compliant
Specifications
Specifications
| Quantity | 1 Ea. |
| Capacity (English) | 17 oz. |
| Product Type | Paint |
| For Use With (Application) | Solvent based Utility, Mark Sites and Stripe Underground Lines, Aisles, Pathways, Parking, Traffic Lines |
| Certifications/Compliance | VOC |
| Color | Fluorescent Orange |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.