Learn More
Accuform Signs Marking Tape Floor Applicator
Catalog No. 19804783 Shop All AccuformNMC Products
Change view
Click to view available options
Product Type:
Marking Tape Floor Applicator
Catalog No. Product Type
19-804-783 Marking Tape Floor Applicator
1 options
Catalog No. 19-804-783 Supplier Accuform Signs Supplier No. HLT300
Add to Cart
Add to Cart
Mark aisles, pathways, and traffic lines by applying tape easily and quickly.
Specifications
For Use With (Equipment) Tape roll size up to 180 ft. L x 4 in. W with 3 in. dia.
Product Type Marking Tape Floor Applicator
For Use With (Application) To mark aisles, pathways, traffic lines by applying tape quickly and easily
Packaging Quantity 1 Ea.