Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Nonpotable Water Labels

Catalog No. 19821783 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
1 x 9 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-821-783 1 x 9 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-821-783 Supplier Accuform Signs Supplier No. LCHL535XVE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use this yellow safety label to alert others that water is not safe for consumption.

Specifications

Material Vinyl
Quantity 1 Ea.
Size (English) 1 x 9 in.
Size (Metric) 2.54 x 22.86 cm
Mounting Type Self-adhesive
For Use With (Application) Designating a Water source, transfer line or storage as unsafe for consumption
Dimensions (L x W) 1 x 9 in.
Displayed Text Non-Potable Water
Product Type Non-potable Water Label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.