Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs S Hooks

Catalog No. 19805123 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-805-123 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-123 Supplier Accuform Signs Supplier No. PRC242YL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-Hooks open ends to quickly link and unlink separate lengths of chain.

  • Works well on plastic stanchion posts

Specifications

Color Yellow
Type S-Hook
For Use With (Application) For linking and unlinking separate lengths of plastic chain
Material Polyethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.