Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Safety Fold-Up Signs: Wet Floor

Catalog No. 19804769 Shop All AccuformNMC Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-804-769 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-769 Supplier Accuform Signs Supplier No. MF111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Free-standing sign alerts of slippery conditions

  • Legend: CAUTION. WET FLOOR (w/ graphic)
  • 0.125 in. high-impact plastic
  • Size (H × W): 50.8 × 30.5cm (20 × 12 in.)
  • Handle cut-out for ease of carrying
  • Folds flat for transport and storage
  • Both sides have the same message
  • Color: Yellow, Black, Red
  • Language: English

Specifications

Material Plastic
Quantity 1 Ea.
For Use With (Application) Identifying temporary hazardous conditions or displaying important messages, quickly and easily
Height (English) 20 in.
Height (Metric) 50.8 cm
Max. Temperature (English) 168°F
Min. Temperature (English) -90°F
Thickness 0.12 in.
Width (English) 12 in.
Width (Metric) 30.5 cm
Temperature Range (Metric) -68°C to +76°C
Temperature Range (English) -90°F to +168°F
Languages English
Displayed Text CAUTION Wet Floor
Product Type Safety Sign
Color Red on Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.