Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Plastic Barricade Tape

Catalog No. 19805303 Shop All AccuformNMC Products
Change view
Click to view available options
Legend: CAUTION BARRICADE TAPE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-805-303 Legend: CAUTION BARRICADE TAPE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-805-303 Supplier Accuform Signs Supplier No. MPT00
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keep employees safe from harm by effectively communicating hazardous situations to others

Barricade tape alerts others of potential hazards in the immediate vicinity.

Specifications

Displayed Text CAUTION
Width (English) 3 in.
For Use With (Application) Ideal for crowd and traffic control purposes
Packaging Quantity 1 Ea.
Thickness 3 mil
Certifications/Compliance ANSI Z535.5-3.2
Color Black on Yellow
Material Polyethylene
Product Type Safety Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.