Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Snap Tite Pipe Markers

Catalog No. 19804877 Shop All AccuformNMC Products
Change view
Click to view available options
Display Text:
Compressed Air
Diameter (English):
0.75 to 1.25 in.
Color:
White, Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text Diameter (English) Color
19-804-877 Compressed Air 0.75 to 1.25 in. White, Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-877 Supplier Accuform Signs Supplier No. RPK227STA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identify contents and mark the directional flow for pipes

Identify contents and indicate the directional flow for pipes carrying compressed air.

Specifications

Description Pipe Marker
Adhesive Type Acrylic
Color White, Blue
Material Vinyl, Plastic
Display Text Compressed Air
For Use With (Application) SNAP' to wrap around and identify contents in pipes with the flow direction
Life Span 1 to 3 Years
Quantity 1 Ea.
Temperature Range (English) -90 to 168°F
Temperature Range (Metric) -67.77 to 75.55°C
Thickness (English) 20 mil
Diameter (English) 0.75 to 1.25 in.
Diameter (Metric) 1.9 to 3.175 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.