Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Style Hasp

Catalog No. 19804740 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-804-740 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-740 Supplier Accuform Signs Supplier No. KDD108
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made from non-sparking style aluminum to avoid potential serious injury to workers

Specifications

Product Type Style Hasps
Color Red
Material Aluminum
Diameter (English) 1 to 1.5 in.
Diameter (Metric) 3 to 4 cm
For Use With (Application) Hazardous environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.