Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform Signs Wash Your Hands Before Leaving Room Signs

Catalog No. 19804822 Shop All AccuformNMC Products
Change view
Click to view available options
Dimensions (L x W):
10 x 14 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-804-822 10 x 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-804-822 Supplier Accuform Signs Supplier No. MRST510VS

missing translation for 'validMainframeMsg'

Notify your team to use proper hygiene procedures in the workplace by washing their hands.

Specifications

Material Vinyl
Quantity 1 Ea.
For Use With (Application) Signs let people employees and people to wash their hands to avoid the spreading of germs
Height (English) 10 in.
Width (English) 14 in.
Dimensions (L x W) 10 x 14 in.
Displayed Text Wash Your Hands Before Leaving Room
Product Type Housekeeping Sign
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.