Learn More
MSC AccuformNMC Hazardous Label, Used Oil Generator, 6x6in Vinyl, 25/Pk.
Catalog No. 5000673752
Encompass
This hazardous material label is designed for Used Oil Generator Information and features fields for shipper, address, source, and contact. It is square, measuring 6 × 6 in, and is made from durable self-laminated vinyl with a removable adhesive.
  • Removable adhesive for flexible application
  • Vinyl material provides durability
  • Graffiti-proof to maintain readability
  • Waterproof for outdoor and wet environments
  • Chemical-resistant for harsh conditions
  • Meets EPA standards for compliance
Catalog No. 50-006-73752 Supplier MSC Supplier No. 56106057
This hazardous material label is designed for Used Oil Generator Information and features fields for shipper, address, source, and contact. It is square, measuring 6 × 6 in, and is made from durable self-laminated vinyl with a removable adhesive.
  • Removable adhesive for flexible application
  • Vinyl material provides durability
  • Graffiti-proof to maintain readability
  • Waterproof for outdoor and wet environments
  • Chemical-resistant for harsh conditions
  • Meets EPA standards for compliance