Learn More
National Diagnostics AccuGel 29:1 (40%), 1 Liter
SDP
Catalog No. 5089990093
Encompass
AccuGel 29:1 is a stabilized, ready-to-use solution of 40% (w/v) acrylamide : bisacrylamide(29:1). AccuGel 29:1 has zero acrylic acid content, eliminating the fixed charges that cause band streaking. Additionally, oxidation products such as aldehydes have been removed by a selective adsorption process. With AccuGel 29:1, you can trust that your results will be consistent from one electrophoretic run to the next. It is certified RNase and DNase Free.
Catalog No. 50-899-90093 Supplier National Diagnostics Supplier No. EC8521LTR
AccuGel 29:1 is a stabilized, ready-to-use solution of 40% (w/v) acrylamide : bisacrylamide(29:1). AccuGel 29:1 has zero acrylic acid content, eliminating the fixed charges that cause band streaking. Additionally, oxidation products such as aldehydes have been removed by a selective adsorption process. With AccuGel 29:1, you can trust that your results will be consistent from one electrophoretic run to the next. It is certified RNase and DNase Free.