Learn More
Biotium AccuGreen™ Broad Range dsDNA Quantitation Kit (500 assays)
Catalog No. 501964579
Encompass_Preferred
The AccuGreen™ Broad Range dsDNA Quantitation Kit is designed for use with handheld fluorometers such as the Qubit fluorometer from Thermo Fisher. The AccuGreen™ kit is specific for dsDNA, and can quantify DNA samples in the range of 0.2-100 ng/uL. The assay has a green fluorescence with an excitation/emission of 500/530 nm (with DNA). This kit has sufficient reagent for 500 assays.
Catalog No. 50-196-4579 Supplier Biotium Supplier No. 31069
The AccuGreen™ Broad Range dsDNA Quantitation Kit is designed for use with handheld fluorometers such as the Qubit fluorometer from Thermo Fisher. The AccuGreen™ kit is specific for dsDNA, and can quantify DNA samples in the range of 0.2-100 ng/uL. The assay has a green fluorescence with an excitation/emission of 500/530 nm (with DNA). This kit has sufficient reagent for 500 assays.