Learn More
Biotium AccuGreen™ Broad Range dsDNA Quantitation Solution (500 assays)
Catalog No. 501964581
Encompass_Preferred
The AccuGreen™ Broad Range dsDNA Quantitation Solution is designed for use with handheld fluorometers such as the Qubit fluorometer from Thermo Fisher. The AccuGreen™ quantitation solution is specific for dsDNA, and can quantify DNA samples in the range of 0.2-100 ng/uL. This item is a standalone fluorescent DNA quantitation solution. The use of this product on the Qubit fluorometer requires two standards. The assay has a green fluorescence with an excitation/emission of 500/530 nm (with DNA). This product has sufficient reagent for 500 assays.
Catalog No. 50-196-4581 Supplier Biotium Supplier No. 31070
The AccuGreen™ Broad Range dsDNA Quantitation Solution is designed for use with handheld fluorometers such as the Qubit fluorometer from Thermo Fisher. The AccuGreen™ quantitation solution is specific for dsDNA, and can quantify DNA samples in the range of 0.2-100 ng/uL. This item is a standalone fluorescent DNA quantitation solution. The use of this product on the Qubit fluorometer requires two standards. The assay has a green fluorescence with an excitation/emission of 500/530 nm (with DNA). This product has sufficient reagent for 500 assays.