Learn More
Biotium AccuGreen™ High Sensitivity dsDNA Quantitation Kit (100 assays)
Catalog No. 501964577
Encompass_Preferred
The AccuGreen™ High Sensitivity dsDNA Quantitation Kit is designed for use with handheld fluorometers such as the Qubit fluorometers from Thermo Fisher. Unlike absorbance-based measurements, AccuGreen™ Dye is highly selective for double-stranded DNA over single-stranded DNA or RNA. The assay has a green fluorescence with an excitation/emission of 502/523 nm (with DNA). The assay has a linear range of 0.1-100 ng dsDNA and a sample concentration range between 5 pg/uL and 100 ng/uL. This trial size kit has sufficient reagent for 100 assays.
Catalog No. 50-196-4577 Supplier Biotium Supplier No. 31066T
The AccuGreen™ High Sensitivity dsDNA Quantitation Kit is designed for use with handheld fluorometers such as the Qubit fluorometers from Thermo Fisher. Unlike absorbance-based measurements, AccuGreen™ Dye is highly selective for double-stranded DNA over single-stranded DNA or RNA. The assay has a green fluorescence with an excitation/emission of 502/523 nm (with DNA). The assay has a linear range of 0.1-100 ng dsDNA and a sample concentration range between 5 pg/uL and 100 ng/uL. This trial size kit has sufficient reagent for 100 assays.