Learn More
Germaine™ Laboratories AccuHome™ Ovulation Test
Catalog No. 23111378
Change view
Click to view available options
Quantity:
1 Test
Catalog No. Quantity
23-111-378 1 Test
1 options
This item is not returnable. View return policy
Catalog No. 23-111-378 Supplier Germaine™ Laboratories Supplier No. 75501
Add to Cart
Add to Cart
This item is not returnable. View return policy
AccuHome™ Ovulation Test detects the LH Surge (Luteinizing Hormone or hLH) in Urine to Aid in the Prediction of Ovulation
Selt test. Results in 5 minutes
Specifications
Content And Storage 15°C to 30°C
CE Marker Yes
Detectable Analytes Luteinizing Hormone Surge (LH or hLH)
Type Urine
Packaging Type Individually wrapped foil pouches
Quantity 1 Test