Learn More
Innovative Cell Technologies Accumax
SDP
Catalog No. NC1738217
Encompass
Accumax is a gentle cell detachment solution containing enzymes that preserve cell surface proteins and receptors. It's suitable for various cell types, ensuring high viability and functionality. Ideal for cell passaging and harvesting, it's available in 100mL and 500mL from Innovative Cell Technologies.
Catalog No. NC1738217 Supplier Innovative Cell Technologies Supplier No. AM105500
Accumax is a gentle cell detachment solution containing enzymes that preserve cell surface proteins and receptors. It's suitable for various cell types, ensuring high viability and functionality. Ideal for cell passaging and harvesting, it's available in 100mL and 500mL from Innovative Cell Technologies.