Learn More
MSC Simple Green Proportioner, Low Flow, 1 GPM, 1 ea
Catalog No. 502868356
Encompass
This proportioner accommodates one product and is designed for low flow applications. It features a single-button operation with a flow rate of 1.0 GPM.
  • Easy product dilution
  • Easy to use
  • Accupro pressure regulation technology
  • Accurate
  • Includes 4 low flow 1 GPM bottle fills, 14 pre-set metering tips, and 1 high flow 3.5 GPM bucket fill output
Catalog No. 50-286-8356 Supplier MSC Supplier No. 41325119
This proportioner accommodates one product and is designed for low flow applications. It features a single-button operation with a flow rate of 1.0 GPM.
  • Easy product dilution
  • Easy to use
  • Accupro pressure regulation technology
  • Accurate
  • Includes 4 low flow 1 GPM bottle fills, 14 pre-set metering tips, and 1 high flow 3.5 GPM bucket fill output