Learn More
STEMCELL Technologies ACCUMAX™, Size: 100 mL
SDP
Catalog No. 501974430
Encompass
ACCUMAX™ is a cell detachment solution of proteolytic and collagenolytic enzymes. It is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue, and the routine detachment of cells from standard tissue culture plasticware and adhesion-coated plasticware. ACCUMAX™ does not contain mammalian- or bacterial-derived products.Each lot of ACCUMAX™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides), and cell detachment from tissue culture plastic.
Catalog No. 50-197-4430 Supplier STEMCELL Technologies Supplier No. 07921
ACCUMAX™ is a cell detachment solution of proteolytic and collagenolytic enzymes. It is useful for creating single-cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue, and the routine detachment of cells from standard tissue culture plasticware and adhesion-coated plasticware. ACCUMAX™ does not contain mammalian- or bacterial-derived products.Each lot of ACCUMAX™ is tested for sterility (by USP membrane filtration method), enzymatic activity (tested with synthetic chromagenic tetrapeptides), and cell detachment from tissue culture plastic.