Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AB200 Benchtop pH/Conductivity Meters

Catalog No. 13636AB200A Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Meter with pH/Cond. Kit
Meter with pH/Cond. Kit, NIST
pH/Conductivity Meter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
13-636-AB200A Meter with pH/Cond. Kit
13-636-AB200 pH/Conductivity Meter
AB200CERT Meter with pH/Cond. Kit, NIST
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-636-AB200A

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Includes

accumet AB200 pH benchtop meter, electrode stand, RS232 and USB cables, 100/240V universal power supply

Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB200 pH/conductivity meter combines the benefits of a pH meter and conductivity meter in one easy to use, reliable measurement system. The large backlit display offers great visibility in laboratory environments.

  • Graphic backlit display with date and time, stability indicator and alarms
  • Dual channel meter for pH and conductivity measurements
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (Cat. No. 13-620-BSP) with five speed settings

NIST-Traceable Calibration Model (-CERT)

  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate

Specifications

  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, 8 Pin DIN, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz

Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial

Certifications

CE, TUV 3-1, FCC Class A

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232 (phono plug), mini-B USB, stirrer
Accuracy (Conductivity) ±1% FS
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Accuracy (Resistivity) ± 1% full scale
Accuracy (Salinity) ± 1% full scale
Accuracy (TDS) ± 1% full scale
Calibration Points (Conductivity) Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (pH) Up to 6 preset or 5 custom
Cell Constant 0.010 to 10.000
Certifications/Compliance Without NIST-traceable pH Certificate
Data Management Details Manual, timed (selectable every 3 to 3600 sec.), printer or CSV format
Data Points 500
Channels 2
Display Type LCD
Measurement Modes pH, mV, Conductivity, TDS, Salinity, Resistivity, Temperature
Range (Conductivity) 0 μS to 500 mS
Range (mV) ±2000 mV
Range (pH) -2 to 20
Range (Resistivity) 2 W to 20 MW
Range (Salinity) 0 to 80 ppt
Range (TDS) 0 ppm to 500 ppt (at TDS factor 1)
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) 0°C to 100°C
Includes Meter, Electrode Arm, RS-232 and USB Cables, 100/240V Power Supply
Inputs BNC, ATC
Model AB200 pH/Conductivity Meter Only
Product Type Meter with pH/Cond. Kit
Resolution (Conductivity) 0.01/0.1μS, 0.001/0.01/0.1mS
Resolution (mV) 0.1mV
Resolution (Resistivity) 0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity) 0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS) 0/0.1ppm, 001/01/0.1ppt
Temperature Accuracy ±0.3°C
Temperature Resolution 0.1°C
Warranty 3 Years
Voltage 100/240 V
Frequency 50/60Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.