Learn More
Fisherbrand™ accumet™ AB315 and AB330 Meter Replacement Universal Power Adapter
Catalog No. 13637010 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
accumet AB315 and AB330 Meters
Catalog No. For Use With (Equipment)
13-637-010 accumet AB315 and AB330 Meters
1 options
Catalog No. 13-637-010
Add to Cart
Add to Cart
Select accessories for your Fisherbrand™ accumet™ AB315 pH/mV meter and Fisherbrand™ accumet™ AB330 conductivity meter.
  • Universal 100/240 V 50/60 Hz power adapter
  • Includes wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement universal power adapter
Frequency 50/60 Hz
Includes Wall plugs for US/Japan, Euro, UK/Singapore, Australia/New Zealand and China
Voltage 100/240 V
Compatibility accumet™ AB300 series meters
For Use With (Equipment) accumet AB315 and AB330 Meters