Learn More
Specifications
Specifications
| Chemical Name or Material | Ion Selective Electrode Reagents |
| Description | Acid Reagent for Residual Chlorine Electrode |
| For Use With (Application) | Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes |
| For Use With (Equipment) | Ion Selective Electrode |
| Quantity | 475 mL |
| Solution Type | Ionic Strength Adjustor |
| Packaging | Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.