Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AE150 Benchtop pH Meter

Catalog No. 13636AE152 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
Bio Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-636-AE152 Bio Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-636-AE152

missing translation for 'validMainframeMsg'

Includes

Double Junction pH Electrode, Temperature Probe and Stand

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price

Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet AE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory

Warranty and Services

3 years on meter

TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ±0.2 ±199.9 ±2 beyond ±200
Outputs Recorder (±2000 mV)
Includes Meter, Double Junction pH Electrode, Temperature Probe and Stand
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV
Calibration Points USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45
Temperature Range (Metric) 0.0°C to 100.0°C
Resolution (Temperature) 0.1°C
Temperature Probe Calibration Offset in 0.1 °C increments; Offset range: ±5 °C
Memory 100 data sets
Inputs BNC, 2.5 mm phono plug, reference pin tip, ground
Warranty 3 Year Meter Warranty
Temperature Compensation Automatic or manual (0 to 100 °C)
Type Bio Set
Electrical Requirements AC/DC 9 V adapter (110/220 VAC, 50-60 Hz)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.