Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

accumet™ Chloride Standard, 100 ppm, Fisherbrand™

Catalog No. 15200931 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-200-931 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-200-931

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Chemical Name or Material Ion Selective Electrode Calibration Standard
Description Chloride Standard, 100 ppm
For Use With (Equipment) Sodium Ion Selective Electrode
Quantity 475 mL
Solution Type Standard
Packaging Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.