Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

accumet™ Cleaning Solution for Oil and Grease Removal, Fisherbrand™

Catalog No. 15200944 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-200-944 4 x 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-200-944

Specifications

Chemical Name or Material pH Electrode Cleaning Solution
Description Cleaning Solution for Oil and Grease Removal
For Use With (Equipment) pH Electrode
Quantity 4 x 60 mL
Solution Type Cleaning
Packaging Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.