Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

accumet™ Fluoride Standard, 1 ppm with TISAB, Fisherbrand™

Catalog No. 15200914 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
475 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-200-914 475 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-200-914
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Ion Selective Electrode Calibration Standard
CAS 7732-18-5, 127-09-3, 7647-14-5, 125572-95-4, 64-19-7, 7681-49-4
Certificate of Analysis Yes
Description Fluoride Standard, 1 ppm with TISAB
For Use With (Application) Calibration
For Use With (Equipment) Fluoride Ion Selective Electrode
Physical Form Liquid
Quantity 475 mL
Solution Type Standard
Packaging Bottle
Traceability to NIST Yes
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.