Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ Glass Body Standard Size Combination Electrodes - Mercury-Free

Catalog No. 13620285 Shop All Fisherbrand Products
Change view
Click to view available options
Connector Types:
BNC
U.S. Standard/Pin
Junction:
Double Ceramic
Single Ceramic
Filling Solution Type:
SP135-500
SP138-500
Length (Metric) Cable:
76.2 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types Junction Filling Solution Type Length (Metric) Cable
13-620-285 BNC Single Ceramic SP135-500 76.2 cm
13-620-223A BNC Double Ceramic SP138-500 76.2 cm
13-620-90 U.S. Standard/Pin Single Ceramic SP135-500 76.2 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 13-620-285
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Removable bulb protector

For routine pH and titration measurements with small samples and small vessels, where separate half-cell electrodes may be impractical

  • 0 to 14 pH range
  • Porous ceramic junction
  • Twist-open/twist-close cap for easy filling
  • Constant distance between FS-5 Universal Glass pH bulb and reference junction means high reproducibility
  • Immersion depth, 4 in. (10.1cm)
  • 30 in. long (76.2cm) leads, except 13-620-91, which has 60 in. long (152.4cm) lead

Choice of Ag/AgCl Single- or Double-Junction internal reference elements

  • Single-junction models with Ag/AgCl reference are perfect for routine pH and titration measurements of aqueous solutions with small samples, small vessels
  • Double-junction models with Ag/AgCl reference are useful in aqueous and nonaqueous solutions, and in applications requiring TRIS buffers
  • For aqueous and nonaqueous samples
  • Double-junction Ag/AgCl eliminate measurement and clogging problems when used in research with Tris buffers
TRUSTED_SUSTAINABILITY

Specifications

Closure Type Twist Open/Twist Close
Color Blue
Connector Types BNC
Junction Single Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 23°C to 212°C
Type Combination pH Electrode
Filling Solution Type SP135-500
For Use With (Application) For routine pH and titration measurements with small samples and small vessels, where separate half cell electrodes may be impractical
For Use With (Equipment) pH meter
Includes Removable bulb protector
Internal Reference Ag/AgCl
Response Time <10 sec.
Temperature (English) Operating -5°F to +100°F
Temperature (Metric) Range -5°C to +100°C
Warranty 1 Year
Length (English) 4 in.
Length (English) Cable 30 in.
Length (Metric) 101.6 mm
Length (Metric) Cable 76.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.