Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

accumet™ TISAB II for Fluoride Electrode, Fisherbrand™

Catalog No. 15200908 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3.8 L
4 x 3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-200-908 3.8 L
15-200-934 4 x 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-200-908
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Ion Selective Electrode Reagents
CAS 7732-18-5, 127-09-3, 7647-14-5, 125572-95-4, 64-19-7
Certificate of Analysis Yes
Description TISAB II for Fluoride Electrode
Concentration 1X
For Use With (Application) Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
For Use With (Equipment) Ion Selective Electrode
pH 5.0 to 5.5 pH
Physical Form Liquid
Quantity 3.8 L
Solution Type Ionic Strength Adjustor
Packaging Gallon
Traceability to NIST Yes
Certifications/Compliance ISO 9001
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.