Learn More
Waters Corp Accumulator Check Valve Assembly, Bio/THF, UPLC System
Catalog No. 502392928 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement accumulator check valve assembly is designed for use with Bio/THF applications. It is specifically utilized on ACQUITY H-Class Bio, ACQUITY Arc Bio, and ACQUITY Premier systems that have been converted to operate with normal phase solvents.
  • Spare parts kits
  • One unit per package
  • Tubing type
  • UPLC system compatibility
  • Annual replacement schedule
Catalog No. 50-239-2928 Supplier Waters Corp Supplier No. 700010388
This replacement accumulator check valve assembly is designed for use with Bio/THF applications. It is specifically utilized on ACQUITY H-Class Bio, ACQUITY Arc Bio, and ACQUITY Premier systems that have been converted to operate with normal phase solvents.
  • Spare parts kits
  • One unit per package
  • Tubing type
  • UPLC system compatibility
  • Annual replacement schedule