Learn More
Biotium AccuOrange™ Protein Quantitation Kit, 200 assays
Catalog No. 501964495
Encompass_Preferred
AccuOrange™ Protein Quantitation Kit is a highly sensitive fluorescence-based assay for quantitating purified protein samples in 96-well format. The detection range of the assay is 0.1-15 ug/mL protein. AccuOrange™ is much more sensitive than traditional protein quantitation assays such as BCA, Bradford and Lowry, and shows superior linearity and reproducibility compared to the NanoOrange protein quantitation assay. The assay shows minimal variability between different proteins, and has stable fluorescence signal for up to 16 hours. The AccuOrange™ assay has low tolerance for non-ionic detergents, and is not recommended for use with cell lysates containing Triton™ X-100, sodium deoxycholate, CHAPs, or other non-ionic detergents. The assay can tolerate up to 0.01% SDS (final concentration in assay). This is a trial size kit and has reagents sufficient for 200 assays in a 96-well microplate format.
Catalog No. 50-196-4495 Supplier Biotium Supplier No. 30071T
AccuOrange™ Protein Quantitation Kit is a highly sensitive fluorescence-based assay for quantitating purified protein samples in 96-well format. The detection range of the assay is 0.1-15 ug/mL protein. AccuOrange™ is much more sensitive than traditional protein quantitation assays such as BCA, Bradford and Lowry, and shows superior linearity and reproducibility compared to the NanoOrange protein quantitation assay. The assay shows minimal variability between different proteins, and has stable fluorescence signal for up to 16 hours. The AccuOrange™ assay has low tolerance for non-ionic detergents, and is not recommended for use with cell lysates containing Triton™ X-100, sodium deoxycholate, CHAPs, or other non-ionic detergents. The assay can tolerate up to 0.01% SDS (final concentration in assay). This is a trial size kit and has reagents sufficient for 200 assays in a 96-well microplate format.