Learn More
Chemglass Life Sciences Chemglass AccuPet Pro Single-Channel Air-Displacement Micropipette, 100-1000 μL
SDP
Catalog No. NC1884661
Encompass_Preferred
The Chemglass Life Sciences AccuPet Pro single-channel micropipette (100-1000 μL) is a research-grade, adjustable air-displacement pipettor designed for routine laboratory liquid handling. It combines ergonomic design with a digital volume indicator and autoclavable components to support accurate, comfortable pipetting during extended use.
  • Ergonomic handle designed to reduce hand and finger fatigue.
  • Digital volume indicator for easy and accurate reading.
  • Continuously adjustable volume control for flexible pipetting.
  • Autoclavable lower portion for convenient sterilization.
  • Research-grade accuracy for consistent liquid handling.
  • Includes tip ejector for safe and convenient tip removal.
Catalog No. NC1884661 Supplier Chemglass Life Sciences Supplier No. CLS3489E07
The Chemglass Life Sciences AccuPet Pro single-channel micropipette (100-1000 μL) is a research-grade, adjustable air-displacement pipettor designed for routine laboratory liquid handling. It combines ergonomic design with a digital volume indicator and autoclavable components to support accurate, comfortable pipetting during extended use.
  • Ergonomic handle designed to reduce hand and finger fatigue.
  • Digital volume indicator for easy and accurate reading.
  • Continuously adjustable volume control for flexible pipetting.
  • Autoclavable lower portion for convenient sterilization.
  • Research-grade accuracy for consistent liquid handling.
  • Includes tip ejector for safe and convenient tip removal.